Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Overnight Express™ Instant TB Medium

Catalog No. 714915 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Each
1 kg
5 Easy
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-491-5 1 kg
71-491-3 1 Each
71-491-4 5 Easy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71-491-5 Supplier MilliporeSigma™ Supplier No. 714915
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as granulated autoinduction culture medium for high-level protein production in pET and other IPTG-inducible bacterial expression systems. Novagen™ Overnight Express™ Instant TB Medium is a granulated medium ensures rapid, uniform dissolution in water, prevents clumping of the medium, and avoids inhalation of airborne powder.

  • Instant TB Medium is combination of Overnight Express Autoinduction System 1 components (Cat. No. 71300) with rich Terrific Broth (TB) medium and is supplied in two formats
  • EasyPak is aluminum foil pouch containing 60g granulated medium, sufficient for 1L culture
  • Add the EasyPak contents to 1L sterile water, supplement with 10mL glycerol, and microwave for 2 min to dissolve
  • Available in 1kg bottles
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Product Type Microbiology Media
For Use With (Application) Protein production
Packaging Type Glass Bottle
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.