Learn More
MilliporeSigma™ Novagen™ PhosphoSafe™ Extraction Reagent
Catalog No. 712964 Shop All MilliporeSigma Products
Catalog No. 71-296-4 Supplier MilliporeSigma™ Supplier No. 712964
Add to Cart
Add to Cart
Extracts cytosolic proteins from mammalian insect cells while preserving phosphorylation state.
EMD Millipore Novagen™ PhosphoSafe™ Extraction Reagent is compatible with kinase assays, protein interaction analysis and other applications.
  • Phosphorylation state preserved during extraction
  • Compatible with kinase assays