Learn More
MilliporeSigma™ Novagen™ pSTBlue-1 AccepTor™ Vector Giga Cloning Kit
Catalog No. 712283 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-228-3 20 Reactions
71-228-4 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-228-3 Supplier MilliporeSigma™ Supplier No. 712283
Add to Cart
Add to Cart
Features NovaBlue GigaSingles™ Competent Cells for increased transformation efficiency. MilliporeSigma™; Novagen™ pSTBlue-1 AccepTor™ Vector Giga Cloning Kit is designed for the cloning of PCR products generated using thermostable DNA polymerases that leave single 3'-dA nucleotide overhangs.
  • Vector enables direct ligation by virtue of single 3'-dU overhangs that anneal with 3'-dA extensions on PCR products, the dU residues are converted to dT residues in vivo
  • No restriction enzymes or special primers
  • Direct ligation of PCR product with vector
  • Compatible with polymerases that leave single 3´-dA overhangs
  • Blue/white screening with pSTBlue-1 or pETBlue™-1 vectors
  • Simple protocol takes as little as 40 minutes from PCR product to plating transformants
TRUSTED_SUSTAINABILITY
Specifications
Product Type pSTBlue-1 Vector Giga Cloning Kit
Content And Storage Store at ≤70°C.
Quantity 20 Reactions