Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ rLysozyme™ Solution

Catalog No. 711105 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1200 kU
300 kU
6000 kU
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-110-5 6000 kU
71-110-4 1200 kU
71-110-3 300 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71-110-5 Supplier MilliporeSigma™ Supplier No. 711105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For lysis of Gram-negative bacteria

  • Contains highly purified and stabilized recombinant lysozyme
  • Catalyzes hydrolysis of N-acetylmuramide linkages in bacterial cell walls
  • Specific activity of rLysozyme (1700KU/mg) for E. coli lysis is 250 times greater than that of chicken egg white lysozyme
  • Optimally active at physiological pH
TRUSTED_SUSTAINABILITY

Specifications

Concentration 27 to 33K U/μL
Components Supplied as a ready-to-use solution at a concentration range of 27 to 33KU/μL in 50% glycerol containing 50mM Tris-HCl, 0.1M NaCl, 0.1mM EDTA, 1 mM DTT, 0.1% TRITON™ X-100
pH 7.5
Content And Storage -20°C
Quantity 6000 kU
Product Type Stabilized recombinant lysozyme

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.