Learn More
MilliporeSigma™ Novagen™ S·Tag™ rEK Purification Kit
Catalog No. 690653 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 U
Catalog No. Quantity
69-065-3 50 U
1 options
Catalog No. 69-065-3 Supplier MilliporeSigma™ Supplier No. 690653
Add to Cart
Add to Cart
Enables target protein release by incubation of the agarose beads with Recombinant Enterokinase (rEK)
  • Enables target protein release by incubation of the agarose beads with Recombinant Enterokinase (rEK), leaving the S•Tag peptide bound to the resin
  • Residual rEK can then be removed efficiently using EKapture Agarose beads
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Multiple storage requirements
Quantity 50 U