Learn More
MilliporeSigma™ Novagen™ Strep·Tactin™ Buffer Kit
Catalog No. 716133 Shop All MilliporeSigma Products
Change view
Click to view available options
Strep·Tactin™ Buffer Kit
Catalog No.
71-613-3 Strep·Tactin™ Buffer Kit
1 options
Catalog No. 71-613-3 Supplier MilliporeSigma™ Supplier No. 716133
Used for purification of recombinant fusion proteins.
  • Pre-tested buffers
  • Convenient, rapid one-step purification of StrepTag™ fusion proteins
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) StrepTactin™ resins
Packaging Glass Bottle
Includes 10X StrepTactin Wash Buffer, 100 mL; 10X StrepTactin Elution Buffer, 25 mL; 10X StrepTactin Regeneration Buffer, 100 mL