Learn More
MilliporeSigma™ Novagen™ Strep·Tactin™ SpinPrep™ Kit
Catalog No. 716083 Shop All MilliporeSigma Products
Change view
Click to view available options
Strep·Tactin™ SpinPrep™ Kit
Catalog No.
71-608-3 Strep·Tactin™ SpinPrep™ Kit
1 options
Catalog No. 71-608-3 Supplier MilliporeSigma™ Supplier No. 716083
Purifies Strep·Tag™ II fusion proteins using small spin columns containing Step·Tactin resin.
  • Each column purifies up to 150μg of Strep·Tag II fusion protein
  • Supplied with 25 Strep·Tactin SpinPrep Columns, receiver tubes, optimized wash and elute buffers