Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ T4 DNA Ligase

Catalog No. 698393 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-839-3 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-839-3 Supplier MilliporeSigma™ Supplier No. 698393
Add to Cart
Add to Cart

For any ligation or cloning application

  • Joins RNA and DNA strands only in duplex molecules and does not join single-stranded nucleic acids
  • Rigorously tested in blue-white cloning assay under conditions that maximize exposure of DNA ends to any contaminating nucleases
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 100 U
Product Type T4 DNA Ligase

For Research use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.