Learn More
MilliporeSigma™ Novagen™ T7·Tag™ Affinity Purification Kit
Catalog No. 690253 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
69-025-3 1 Kit
1 options
Catalog No. 69-025-3 Supplier MilliporeSigma™ Supplier No. 690253
Add to Cart
Add to Cart
Rapid immunoaffinity purification of target proteins that carry the T7·Tag sequence
  • Designed for rapid immunoaffinity purification of target proteins that carry the T7•Tag sequence
  • Purification is based on binding target proteins to T7•Tag Monoclonal Antibody that is covalently coupled to cross-linked agarose beads, washing away unbound proteins, and eluting at pH 2.2
  • Neutralization Buffer is included to limit protein exposure to low pH
  • Capacity will vary somewhat between different target proteins, but the beads are standardized to bind a minimum of 300μg T7•Tag β-galactosidase per milliliter of settled resin
  • The beads can be used in either batch or column methods and can be regenerated a minimum of five times without loss of binding activity
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at +2°C to +8°C
Quantity 1 Kit