Description
EMD Millipore Novagen™ T7 .Tag Antibody Agarose is based on binding target proteins to T7 Tag monoclonal antibody that is covalently coupled to cross-linked agarose beads.
- Washes unbound proteins, elutes at pH 2.2
- Beads are standardized to bind minimum of 300μg T7 Tag β-galactosidase per mm of settled resin
- Beads can be used in either batch or column methods
- Recycled minimum of five times without loss of binding activity
Specifications
Specifications
| Quantity | 2 mL |
| Storage Requirements | 2° to 8°C |
| Product Type | Agarose Gel |