Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Terrific Broth

Catalog No. 717545 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
5 kg
10 kg
Packaging Type:
Glass Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
71-754-5 10 kg Plastic Drum
71-754-3 500 g Glass Bottle
71-754-4 5 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71-754-5 Supplier MilliporeSigma™ Supplier No. 717545
Add to Cart
Add to Cart

Used as highly enriched granulated medium to improve the yield of plasmid DNA from E. coli. MilliporeSigma™; Novagen™ Terrific Broth granules ensure rapid and uniform dissolution in water.

  • Prevents clumping of medium and inhalation of airborne powder
  • Composition of the Terrific Broth per liter is 12g tryptone, 24g yeast extract, 9.4g potassium phosphate, dibasic, and 2.2g potassium phosphate, monobasic
  • Use 47.6g of granulated medium per liter of water
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Product Type Microbiology Media
Packaging Type Plastic Drum
Quantity 10 kg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.