Learn More
AdipoGen Novaluron
SDP
Supplier:  AdipoGen CDXN0069M500
Encompass_Preferred
Chemical. CAS 116714-46-6. Formula C17H9ClF8N2O4. MW 492.7. Synthetic. Acylurea insecticide. Insect growth inhibitor. Chitin synthase inhibitor active against larvae of Coleoptera, Homoptera, Diptera and Lepidoptera. Shown to be an inhibitor of acetylcholinesterase in B-biotype Bemisia tabaci. Compound can be used as analytical reference material.
Catalog No. 50-256-0417
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.