Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ NovaShield HRP Stabilizer (Carrier-Free)
SDP

Catalog No. NBP261912 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61912 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61912 Supplier Novus Biologicals™ Supplier No. NBP261912

Ready-to-use Peroxidase (HRP) Conjugate Stabilizer.

  • Superior HRP protection
  • Preserves and stabilizes the activity of HRP conjugates, enzymes and other bio-molecules used in immunoassays and molecular assays
  • Minimizes background interactions and issues
  • Stable through freeze-thaws
  • Does not contain mammalian proteins, bovine proteins, or BSA (Carrier-Free)

Specifications

Content And Storage Store at 4°C in the dark.
Quantity 200 mL
Target NovaShield HRP Stabilizer (Carrier-Free)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.