Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ NOVAX™ Class 3 Black Natural Rubber Electrical Insulating Gloves

Catalog No. 19100545 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
9
10
11
12
Length (English):
16 in.
18 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English)
19-100-545 9 18 in.
19-100-541 10 16 in.
19-100-546 10 18 in.
19-100-542 11 16 in.
19-100-547 11 18 in.
19-100-543 12 16 in.
19-100-548 12 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-100-545 Supplier PIP™ Supplier No. 155318/9
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves are constructed of natural rubber to provide dielectric protection, and compliant with ANSI/ASTM D120 and NFPA 70E standards for use around electrical hazards.

  • Class 3 electrical insulating gloves constructed of natural rubber
  • Flexible material allows workers to easily handle small, delicate parts
  • Contoured shape is comfortable and helps to reduce hand fatigue
  • Gloves meet or exceed all requirements of ANSI/ASTM D120 and NFPA 70E
  • Ideal for use around electrical and arc flash hazards
  • Individually numbered for traceability
  • Stamped with factory manufacturing date
  • Silicone-free

Specifications

Certifications/Compliance ANSI/ASTM D120, Class 3, Type I, NFPA 70E
Applications Industrial
Color Black, Orange
Exterior Finish No Texture
Hand Specific
Material Natural Rubber
Product Type Lineman's Gloves
Size 9
For Use With (Application) Electrical Utility Work
Glove Cuff Style Straight
Series 155-3-18
Length (English) 18 in.
Length (Metric) 45.7 cm
Voltage 26,500 V
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.