Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ NOVAX™ Class 0 Electrical Safety Gloves Kit

Catalog No. 191682132 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Black
Orange
Size:
8
9
10
11
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-168-2132 Black 8
19-168-2133 Black 9
19-168-2134 Black 10
19-168-2135 Black 11
19-168-2100 Orange 8
19-168-2101 Orange 9
19-168-2102 Orange 10
19-168-2103 Orange 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-168-2132 Supplier PIP™ Supplier No. 150SK0/8KIT
Add to Cart
Add to Cart
This item is not returnable. View return policy

Kit includes one pair of natural rubber electrical insulating gloves, a pair of top grain goatskin leather glove protectors (148-1000), and an 11 in. nylon glove bag.

  • Includes one pair of natural rubber electrical insulating gloves, a pair of top grain goatskin leather glove protectors (148-1000), and an 11 in. nylon glove bag
  • Class 0 insulating gloves will protect workers from up to 1000 V
  • Meets NFPA 70E standards for use around arc flash hazards

Specifications

Certifications/Compliance ANSI/ASTM D120, Class 0, Type I, NFPA 70E
Applications Industrial
Color Black
Hand Specific
Material Natural Rubber, Top Grain Goatskin Leather
Product Type Electrical Insulating Gloves Kit
Size 8
For Use With (Application) Electrical Utility Work, Industrial Plant Maintenance, and Power Generation
Glove Cuff Style Straight
Includes Natural Rubber Electrical Insulating Gloves (150-0-11), Top Grain Goatskin Leather Glove Protectors (148-1000), and a Nylon Glove Bag (148-2136)
Series 150-SK-0
Length (English) 11 in.
Length (Metric) 28 cm
Voltage 1000 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.