Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ NOVAX™ Class 2 Electrical Safety Gloves Kit

Catalog No. 191682140 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Black
Orange
Size:
8
9
10
11
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-168-2140 Black 8
19-168-2141 Black 9
19-168-2142 Black 10
19-168-2143 Black 11
19-168-2108 Orange 8
19-168-2109 Orange 9
19-168-2110 Orange 10
19-168-2111 Orange 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-168-2140 Supplier PIP™ Supplier No. 150SK2/8KIT
Add to Cart
Add to Cart
This item is not returnable. View return policy

Kit includes one pair of natural rubber electrical insulating gloves (150-2-14), a pair of cowhide leather glove protectors (148-4000), and a 14 in. nylon glove bag.

  • Includes one pair of natural rubber electrical insulating gloves, a pair of top grain cowhide leather glove protectors (148-4000), and an 14 in. nylon glove bag
  • Class 2 insulating gloves will protect workers from up to 17,000 V
  • Meets NFPA 70E standards for use around arc flash hazards

Specifications

Certifications/Compliance ANSI/ASTM D120, Class 2, Type I, NFPA 70E
Applications Industrial
Color Black
Hand Specific
Material Natural Rubber, Top Grain Cowhide Leather
Product Type Electrical Insulating Gloves Kit
Size 8
For Use With (Application) Electrical Utility Work, Industrial Plant Maintenance, and Power Generation
Glove Cuff Style Straight
Includes Natural Rubber Electrical Insulating Gloves (150-2-14), Top Grain Cowhide Leather Glove Protectors (148-4000), and a Nylon Storage Bag (148-2142)
Series 150-SK-2
Length (English) 14 in.
Length (Metric) 35.5 cm
Voltage 17,000 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.