Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novec™ 7100 Engineered Fluid, Sigma-Aldrich

Catalog No. 01258823 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-823 1 L
01-258-913 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-823 Supplier MilliporeSigma Supelco Supplier No. SHH00021L
Add to Cart
Add to Cart
  • Cleaning and rinsing agent
    • Heavy-duty cleaning (co-solvent) - heavy oils, greases, fluxes
    • Medium-duty cleaning (azeotrope) - oils, greases, waxes
    • Light-duty cleaning (neat) - particulates, fluorolubes, light oils, fluoropolymer
  • Lubricant carrier: fluorocarbons, hydrocarbons, and silicones
  • Spot-free water drying agent (with surfactants added)
  • Specialty solvents, dispersion media, reaction media
  • Spray contact cleaner
  • CFC, HCFC, HFC and PFC replacement
  • Dielectric test media
  • Heat transfer
  • Preservation of biological specimens
  • Cryofreezing solution
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163702-07-6
Molecular Weight (g/mol) 250.06 g/mol

Specifications

Quantity 1 L
Packaging Amber Bottle
Percent Purity ≥99.5%
Chemical Name or Material Methoxy-nonafluorobutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.