Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Amyloid Precursor Protein ELISA Kit

Catalog No. KHB0051
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KHB0051 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KHB0051 Supplier Invitrogen™ Supplier No. KHB0051
Only null left
Add to Cart
Add to Cart

ELISA

The Human Amyloid Precursor Protein (APP) ELISA quantitates Hu APP in human cerebrospinal fluid and cell lysates. The assay will exclusively recognize both natural and recombinant Hu APP. Principle of the method The Human APP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Amyloid Precursor Protein (APP) or Amyloid beta precursor protein functions as a cell surface kinesin I membrane receptor, mediating the axonal transport of beta-secretase and presenilin 1. APP is important for neurite growth, neuronal adhesion and axonogenesis. APP is a 100-140 kDa transmembrane glycoprotein that exists as several isoforms resulting from alternative splicing. Proteolytic cleavage of APP by beta- and gamma-secretases results in the generation of beta amyloid, which is the primary component of senile plaques. Senile plaques are one of the major histopathologic features of Alzheimer's disease. Abnormal regulation and processing of APP also plays a role in Down's syndrome, early onset familial Alzheimer's disease, and cerebral hemorrhage.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05067
Assay Range 0.78 to 50 ng/mL
Assay Sensitivity <0.4 ng/mL
Concentration 0.78 to 50 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates, Cerebrospinal Fluid
Substrate Type Amyloid-Related Proteins
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 351
Gene Symbol AAA,ABETA,ABPP,AD1I,CTFgamma,CVAP,PN-II,PN2,APP
Incubation Time 4 hr.
Interassay CV 7.4%
Intraassay CV 6.3
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene APP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 10 μL, Cerebrospinal Fluid 10 μL
Storage Requirements 2°C to 8°C
Target Amyloid Precursor Protein
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.