Learn More
Invitrogen™ Mini Blot Module Gasket
Catalog No. B1001
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 unit
Catalog No. Quantity
B1001 1 unit
1 options
Catalog No. B1001 Supplier Invitrogen™ Supplier No. B1001
Add to Cart
Add to Cart
For Bolt™ Mini Blot Module
Gasket fits on the cathode core (-) of the blot module. Tweezers designed for handling nitrocellulose or PVDF membranes.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Bolt™
Gel Size Mini
Product Line Bolt
Type Gasket
Quantity 1 unit
For Research Use Only. Not for use in diagnostic procedures.