Learn More
Invitrogen™ Blotting Tweezer
Catalog No. B1002
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 unit
Catalog No. Quantity
B1002 1 unit
1 options
Catalog No. B1002 Supplier Invitrogen™ Supplier No. B1002
  • $28.70
    $27.65 / Each of 1
    Save $1.05
    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
For Bolt™ Mini Blot Module
Gasket fits on the cathode core (-) of the blot module. Tweezers designed for handling nitrocellulose or PVDF membranes.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Protein assays and analysis, protein gel electrophoresis
For Use With (Equipment) Bolt Mini Blot Module
Gel Size Mini
Type Tweezers
Quantity 1 unit
For Research Use Only. Not for use in diagnostic procedures.