Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gel Runner Tank

Catalog No. B4478641
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4478641 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4478641 Supplier Invitrogen™ Supplier No. B4478641
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For the Bolt Mini Gel Tank

The Bolt Tank Lid fits the top of the Gel Runner Tank. The Cassette Clamps (right and left) lock and seal the gel cassettes within the Gel Runner Tank, and the orientation is determined when facing the front of the gel tank. The Bolt Tank Base holds the Gel Runner Tank. The replacement Gel Runner Tank does not include tank lid, tank base, or cassette clamps.
The Bolt Mini Gel Tank is compatible with Bolt Bis-Tris Plus gels, and can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.

Bolt Bis-Tris Plus Gels

  • Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of small- to medium-sized proteins under denaturing conditions
  • Similar to NuPAGE™ Bis-Tris gels, Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications
  • Bolt gels are ideal for Western blot transfer and analysis, and any other techniques where protein integrity is crucial; also use Bolt gels to obtain optimal results for day-to-day protein separation needs
  • Bolt Bis-Tris Plus gels come in four gel types and multiple well formats

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Runner Tank
For Use With (Equipment) Bolt™ System
Gel Compatibility Bolt™ Bis-Tris Plus Gels
Gel Size Mini
Product Line Bolt
Type Gel Runner Tank
Quantity 1 unit
Can I purchase replacement parts for the Mini Blot Module?

Here are the replacement parts we offer for the Mini Blot Module:

- Mini Blot Module Gasket, Cat. No. B1001
- Gel Knife, Cat. No. EI9010
- Bolt Tank Base, Cat. No. B4478640
- Gel Runner Tank, Cat. No. B4478641
- Mini Gel Tank Lid, Cat. No. A25944
- Cassette Clamp, Right, Cat. No. B4478592
- Cassette Clamp, Left, Cat. No. B4478593
- Power Supply Adapters, Cat. No. ZA10001

Can I purchase replacement parts for the Mini Gel Tank?

Here are the replacement parts we offer for the Mini Gel Tank:
Replacement part - Cat. No.
Gel Runner Tank - B4478641
Mini Gel Tank Lid - A25944
Mini Gel Tank Base - A25950
Cassette Clamp, left - A25946
Cassette Clamp, right - A25945
Cassette Clamp Cam Handle Set - A26732

What are the components of the Mini Gel Tank?

The Mini Gel Tank includes the Mini Gel Electrophoresis Tank, Mini Gel Tank Lid, Cassette Clamp (Right), 10 cm, Cassette Clamp (Left), 10 cm, Mini Gel Tank Base, Invitrogen Power Supply Adapters, and Gel Knife.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.