Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NativePAGE™ 5% G-250 Sample Additive

Catalog No. BN2004
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BN2004 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BN2004 Supplier Invitrogen™ Supplier No. BN2004
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Add to samples prepared with the NativePAGE Sample Buffer and detergents (10% DMM or 5% Digitonin) just prior to applying the samples to NativePAGE gels

This Additive is also included in the NativePAGE Sample Prep Kit. The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE Sample Prep Kit are compatible with NativePAGE Novex Bis-Tris Gels for native electrophoresis showing increased resolution and reduced streaking.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Buffer Sample Loading Buffers
Gel Type Native-PAGE
Gel Compatibility NativePAGE™ Gels
Quantity 0.5 mL
Shipping Condition Room Temperature
Product Line NativePAGE
Product Type PAGE G-250 Sample Additive
I would like to run a NativePAGE gel. Which of your protein standards should I use?

We recommend using the NativeMark Unstained Protein Standard, Cat. No. LC0725 for native gel electrophoresis with Tris-Glycine, NuPAGE Tris-Acetate or NativePAGE gels.

Can I use the NativePAGE Sample and Running buffers with NuPAGE Bis-Tris gels?

We do not recommend using NativePAGE Sample and Running buffers with NuPAGE Bis-Tris gels. NuPAGE Invitrogen Bis-Tris gels are optimized for denaturing conditions and have an extremely low operating pH (pH 7.0), which makes it difficult for most proteins to migrate through them in a native state.

Can I use the NativePAGE Sample and Running buffers with Invitrogen Tris-Glycine gels or NuPAGE Tris-Acetate gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate or Invitrogen Tris-Glycine gels. For those gels, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.

What is the purpose of using the NativePAGE 5% G-250 Sample Additive?

The NativePAGE 5% G-250 Sample Additive is a concentrated stock solution of Coomassie G-250 designed for use with detergent (non-ionic) containing samples prepared for NativePAGE gel electrophoresis. Native proteins normally migrate on the gel according to natural charge/pI in the pH of gel buffer, but adding the Coomassie G-250 renders an associated negative charge even to high pI proteins that would normally have a positive charge. It binds to proteins non-specifically without denaturing them. The NativePAGE 5% G-250 Sample Additive is added to the cathode buffer and, optionally, to samples as well.

What is the composition of the NativePAGE 5% G-250 Sample Additive? Does it contain a detergent?

The NativePAGE 5% G-250 Sample Additive (Cat. No. BN2004) is a concentrated stock solution of Coomassie G-250 designed for use with detergent (non-ionic) containing samples prepared for NativePAGE gel electrophoresis. The exact composition of the NativePAGE 5% G-250 Sample Additive is proprietary. It does not contain a detergent.

Can I use the NativePAGE Sample and Running buffers with Invitrogen Tris-Glycine gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including Invitrogen Tris-Glycine gels. For those gels, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.

Can I use the NativePAGE Sample and Running buffers with NuPAGE Tris-Acetate gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate gels. When running Tris-Acetate gels under native conditions, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.