Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IR (Phospho) [pY1162/pY1163] ELISA Kit

Catalog No. KHR9131
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KHR9131 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KHR9131 Supplier Invitrogen™ Supplier No. KHR9131
Add to Cart
Add to Cart

ELISA

Kit is designed to detect and quantify the level of IR (Phospho) [pY1162/pY1163] in fresh or frozen human cell lysates. Cross-reactivity with mouse and rat cells has been observed. The assay recognizes both natural and recomb. Human IR (Phospho) [pY1162/pY1163]. Principle of the method A monoclonal capture antibody specific for IR (Phospho) [pY1162/pY1163] has been coated onto the wells of 96-well plate. During the first incubation, standards of known content and unknown samples are pipetted into the wells and the antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for the target protein is added to the wells and serves as a detection antibody by binding to the immobilized protein captured during the first incubation. After washing, a HRP labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of target protein present in the original specimen and the optical density can be read on a standard microplate reader. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

INSR is a receptor tyrosine kinase. Binding of insulin to the extracellular domain of INSR stimulates glucose uptake. INSR and IGF-1 receptors share major structural and functional similarity. The earliest cellular response to insulin stimulation is autophosphorylation of tyrosine in INSR. In humans, the INSR gene is located on chromosome 19. Defects in INSR are the cause of various insulin resistance syndromes and IGF-1R defects may also cause some forms of growth retardation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06213,P15127,P15208
Assay Range 0.625 to 40 U/mL
Assay Sensitivity <0.4 U/mL
Concentration 1.6 U/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Substrate Type Receptor Tyrosine Kinases
Target Species Mouse, Rat
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 16337, 24954, 3643
Gene Symbol CD220,HHF5
Incubation Time 4 hr.
Interassay CV 7.6%
Intraassay CV 6
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene IR
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 10 μL
Storage Requirements 2°C to 8°C
Target INSR
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.