Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human JAK2 (Total) ELISA Kit

Catalog No. KHO5521
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KHO5521 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KHO5521 Supplier Invitrogen™ Supplier No. KHO5521
Add to Cart
Add to Cart

ELISA

The Human JAK2 (Total)) ELISA Kit is designed to detect and quantify the level of JAK2 (Total) in fresh or frozen human or mouse cell lysates. The assay recognizes both natural and recombinant human JAK2 (Total), independent of its phosphorylation state. Principle of the method A monoclonal capture antibody specific for JAK2 (Total) has been coated onto the wells of the 96-well plate. During the first incubation, standards of known content and unknown samples are pipetted into the wells and the antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for the target protein is added to the wells and serves as a detection antibody by binding to the immobilized protein captured during the first incubation. After washing, a horseradish peroxidase labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of target protein present in the original specimen and the optical density can be read on a standard microplate reader. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Janus kinase 2 (JAK2) is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways and is mutated in polycythemia vera. This kinase contains the catalytic domain (JH1) of JAK2. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex. Mutations affecting JAK2 can cause Budd-Chiari syndrome, Polycythemia vera, Thrombocythemia 3, myelofibrosis, or Acute myelogenous leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60674,Q62120
Assay Range 0.95 to 60 ng/mL
Assay Sensitivity <0.95 ng/mL
Concentration 0.95 to 60 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Substrate Type Cytoplasmic Tyrosine Kinases
Target Species Mouse, Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 16452, 3717
Gene Symbol JTK10,THCYT3
Incubation Time 4 hr.
Interassay CV 8.1%
Intraassay CV 5.1
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene JAK2
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 10 μL
Storage Requirements 2°C to 8°C
Target JAK2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.