Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUMO Protease

Catalog No. 12588018
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-588-018 250 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-588-018 Supplier Invitrogen™ Supplier No. 12588018
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

SUMO Protease is provided with 10X high salt and 10X no salt dilution buffers.

A recombinant fragment of ULP1 (Ubl-specific protease 1) from Saccharomyces cerevisiae

SUMO Protease, also known as Ulp, is highly specific for the SUMO protein fusion, recognizing the tertiary structure of SUMO rather than an amino acid sequence.

  • Highly active cleavage
  • No non-specific proteolysis (highly specific cleavage)
  • Activity from 2°C to 37°
  • A six-histidine sequence to facilitate its removal from the digested protein sample

Application:Removal of fusion tags from recombinant proteins
Unit Definition: One unit of SUMO Protease is defined as the amount of enzyme needed to cleave 85% of 2μg of substrate protein at 30°C in one hour
Quality Control: SUMO Protease has greater than 85% single-band purity with no non-specific protease contamination; functionally tested for the absence of any non-specific protease activity

GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Host Species S. cerevisiae
Content And Storage SUMO Protease is provided with 10X high salt and 10X no salt dilution buffers. Store at -80°C. Guaranteed stable for 6 months when properly stored.
Shipping Condition Dry Ice
Quantity 250 units
Are NP40 and DTT required for SUMO Protease activity? Will they interfere with downstream applications?

Yes, they are both required, although it might be possible to substitute the NP40 with another detergent via dialysis if you have concern about interference with mass spectrometry. DTT is an absolute requirement.

What enzyme should I choose if I do not want any extra amino acids left on my protein of interest after cleavage?

We would recommend using our SUMO system.

How does SUMO protease work?

SUMO protease, also known as Ulp, is a recombinant fragment of ULP1 (Ubl-specific protease 1) from Saccharomyces cerevisiae. It is highly specific for the SUMO protein fusion, recognizing the tertiary structure of SUMO rather than an amino acid sequence. The SUMO protease itself has a His tag for easy removal from the protein mix after cleavage.

What products do you offer for enzymatic cleavage of fusion tags from recombinant proteins?

We offer the following products:

-AcTEV Protease (Cat. Nos. 12575015, 12575023)
-EKMax Enterokinase (Cat. Nos. E18001, E18002)
-SUMO Protease (Cat. No. 12588018)

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.