Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ Tris-Glycine SDS Buffer Kit

Catalog No. LC2677
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC2677 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC2677 Supplier Invitrogen™ Supplier No. LC2677
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.

Made with high-purity reagents and are strictly quality controlled

Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Product Line Novex
Gel Compatibility Novex Tris-Glycine Gel
Product Type Buffer Kit
Gel Type SDS PAGE Gel
Content And Storage • 500 mL 10X Tris-Glycine SDS Running Buffer
• 20 mL 2X Tris-Glycine SDS Sample Buffer

All Novex™ buffers have a 6 month shelf life. Store at +4°C to 25°C, depending upon formulation.
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.