Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 12%

Catalog No. STM5015
Encompass_Preferred
Change view
Click to view available options
No. of Wells:
20-well
26-well
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
STM5015 26-well
STM5014 20-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. STM5015 Supplier Invitrogen™ Supplier No. STM5015
Get Up to 55% Off Protein Gel Welcome Packs
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Novex Tris-Glycine Plus Midi Welcome Pack, Wedgewell format provides all the necessary Novex Tris-Glycine Plus Midi gels, buffers, and reagents you will need to begin using the SureLock Tandem Midi Gel Tank.

The Novex Tris-Glycine Plus Midi Wedgewell Welcome Pack provides all the necessary Novex Tris-Glycine Plus Midi gels, buffers, and reagents you will need to begin using the SureLock Tandem Midi Gel Tank.

Welcome Pack contents

  • SureLock Tandem Midi Gel Tank (STM1001)
  • Novex Tris-Glycine Plus WedgeWell Midi Gels (2 boxes, 20 gels)
  • Novex Tris-Glycine SDS Running Buffer, 10X (LC2675)
  • Novex Tris-Glycine SDS Sample Buffer, 2X (LC2676)
  • NuPAGE Sample Reducing Agent, 10X (NP0004)
  • PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the SureLock Tandem Midi Gel Tank

The SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a set-up time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.

About Novex Tris-Glycine Plus Midi gels

Novex Tris-Glycine Plus midi gels are polyacrylamide gels based on traditional Laemmli protein electrophoresis, allowing the use of Laemmli sample and running buffers. The gels offer reproducible separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine Plus midi gels are a 1.0-mm-thick, wider-format (8 cm x 13 cm) gel for higher throughput electrophoresis of protein samples.

Transfer and detection

For transfer of proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) for traditional wet transfer using the SureLock Tandem Midi Blot Module (STM2001). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Convenient to use

Novex Tris-Glycine WedgeWell-format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) SureLock Tandem Midi Gel Tank
Gel Size Midi
Product Line Novex
Gel Percentage 12%
Gel Type Tris-Glycine Plus
No. of Wells 26-well
Quantity 1 Welcome Pk.
Separation Range 20 to 250 kDa
Shipping Condition Wet Ice
Thickness (Metric) 1.0 mm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.