Learn More
Invitrogen™ Novex™ WedgeWell™ Tris-Glycine Welcome Pack, 4 to 20%
Description
The Novex Tris-Glycine Welcome Pack provides all of the necessary Novex Tris-glycine gels, buffers, and reagents needed to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Novex and NuPAGE mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.
The Novex Tris-Glycine Welcome Pack contains:
• Mini Gel Tank (A25977)
• Novex Tris-Glycine Mini Gels, WedgeWell format (2 boxes, 20 gels)
• Novex Tris-Glycine SDS Running Buffer, 10X (LC2675)
• Novex Tris-Glycine SDS Sample Buffer, 2X (LC2676)
• NuPAGE Sample Reducing Agent, 10X (NP0004)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)
About the Mini Gel Tank
The Mini Gel Tank is compatible with all Novex and NuPAGE mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.
About Novex Tris-Glycine gels, WedgeWell format
Novex Tris-Glycine mini gels are polyacrylamide gels based on traditional Laemmli protein electrophoresis, allowing the use of Laemmli sample and running buffers. They enable consistent performance and separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.
For transferring your proteins to a membrane, we recommend using for Novex Tris-Glycine Native Running Buffer (LC2672) or Novex Tris-Glycine Transfer Buffer (LC3675) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).
Specifications
Specifications
| Gel Percentage | 4 to 20% |
| Gel Size | Mini |
| Gel Thickness | 1.0 mm |
| Gel Type | Tris-Glycine |
| Separation Range | 20 to 200 kDa |
| Wells | 12-well |
| Quantity | 1 Welcome Pk. kit |
| Shipping Condition | Wet Ice |
| Product Line | Novex, WedgeWell |
| For Use With (Equipment) | Mini Gel Tank |
Frequently Asked Questions (FAQs)
No. Novex WedgeWell gels must be stored at 4 degrees C.
The Novex WedgeWell gels can use standard Tris-Glycine sample and running buffers. For running under native conditions, we recommend using sample and running buffers that do not contain SDS (such as our Native Tris-Glycine premade buffers).
We were able to optimize shelf life through a proprietary gel formulation change. Unfortunately, we are unable to provide specific details on the chemical changes.
We use what we call mirror printing', the same way we do for Bolt gels. This means that when the gel is loaded in the Mini Gel Tank, the text will face in the readable direction (left to right).
Yes, the new Novex WedgeWell gels can be run in the XCell SureLock Mini-Cell.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.