Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ Zymogram Renaturing Buffer (10X)

Catalog No. LC2670
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC2670 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC2670 Supplier Invitrogen™ Supplier No. LC2670
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Used with Novex™ Zymogram Gels

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Buffer Renaturing Buffers
Concentration 10 X
Gel Type Zymogram
Gel Compatibility Novex™ Zymogram Gels
Quantity 500 mL
Shipping Condition Room Temperature
Product Line Novex
Product Type Zymogram Renaturing Buffer
Does the Colloidal Blue Staining kit work with Invitrogen Zymogram gels?

Yes, the Colloidal Blue Staining kit works very well with Invitrogen Zymogram gels.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.