Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novobiocin sodium salt

Catalog No. AAJ6092806
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6092806 5 g
AAJ6092803 1 g
AAJ6092809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6092806 Supplier Thermo Scientific Chemicals Supplier No. J6092806
Add to Cart

Novobiocin sodium salt, CAS # 1476-53-5, is a topoisomerase II inhibitor capable of inhibiting DNA synthesis. This compound is produced by Streptomyces nieveus and shows antibiotic activity. | CAS: 1476-53-5 | C31H35N2NaO11 | 634.61 g/mol

Topoisomerase II inhibitorNovobiocin sodium salt is effective an antistaphylococcal agent used in the treatment of Methicillin-resistant Staphylococcus aureus (MRSA). It is also used to inhibit the activity of retroviral RNA-dependent DNA polymerase as well as inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Novobiocin sodium salt, first isolated from Streptomyces nieveus, is a topoisomerase II inhibitor. It is a coumarin compound showing antibiotic activity
• Novobiocin sodium salt can inhibit DNA synthesis by inhibiting the topoisomerase II enzyme. Its antibacterial activity arises from competitive inhibition of the ATPase reaction catalyzed by the GyrB subunit

Application

• Novobiocin sodium salt is used to inhibit the activity of retroviral RNA-dependent DNA polymerase in laboratory experiments
• It is also used as an inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase.
• Novobiocin sodium salt shows anti-staphylococcal activity against methicillin-resistant Staphylococcus aureus, a gram-positive bacterium
• It permits the relaxation of negative DNA supercoils and has been used to generate positively supercoiled DNA
• This compound inhibits the chaperone activity of Hsp90

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1476-53-5
Molecular Formula C31H35N2NaO11
Molecular Weight (g/mol) 634.61
MDL Number MFCD00066541,MFCD00066541
InChI Key AXOUUAINTJNFRS-UHFFFAOYNA-N
PubChem CID 131673945
IUPAC Name sodium 7-{[4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy}-3-[4-hydroxy-3-(3-methylbut-2-en-1-yl)benzamido]-8-methyl-2,4-dioxo-3,4-dihydro-2H-1-benzopyran-3-ide
SMILES [Na+].COC1C(OC(N)=O)C(O)C(OC2=CC=C3C(=O)[C-](NC(=O)C4=CC=C(O)C(CC=C(C)C)=C4)C(=O)OC3=C2C)OC1(C)C

Specifications

Type Novobiocin Sodium Salt
Physical Form Powder
Melting Point 215°C
Color Brown to White
Quantity 5 g
Beilstein 3892910
Merck Index 14,6722
Solubility Information Soluble in water at 100mg/ml; Also soluble in ethanol or DMSOSoluble in water,ethanol,dimethyldulfoxide and methanol.
Formula Weight 634.61
Chemical Name or Material Novobiocin Sodium Salt
Hazard Category Warning
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash contaminated clothing before reuse.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 216-023-6
RTECSNumber RD5425000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.