Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ACAA1 Protein
SDP

Catalog No. p-5354315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14532301 0.1 mg
NBP1453235 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14532301 Supplier Novus Biologicals™ Supplier No. NBP1453230.1MG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 27-424 of Human ACAA1 The Recombinant Human ACAA1 Protein is derived from E. coli. The Recombinant Human ACAA1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1.0mg/mL
For Use With (Application) ELISA, Western Blot
Formulation In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol 1mM DTT, 0.1M NaCl
Gene ID (Entrez) 30
Molecular Weight (g/mol) 43.8kDa
Purification Method Protein
Quantity 0.1 mg
Source Human
Immunogen MGSSHHHHHH SSGLVPRGSH MLSGAPQASA ADVVVVHGRR TAICRAGRGG FKDTTPDELL SAVMTAVLKD VNLRPEQLGD ICVGNVLQPG AGAIMARIAQ FLSDIPETVP LSTVNRQCSS GLQAVASIAG GIRNGSYDIG MACGVESMSL ADRGNPGNIT SRLMEKEKAR DCLIPMGITS ENVAERFGIS REKQDTFALA SQQKAARAQS KGCFQAEIVP VTTTVHDDKG TKRSITVTQD EGIRPSTTME GLAKLKPAFK KDGSTTAGNS SQVSDGAAAI LLARRSKAEE LGLPILGVLR SYAVVGVPPD IMGIGPAYAI PVALQKAGLT VSDVDIFEIN EAFASQAAYC VEKLRLPPEK VNPLGGAVAL GHPLGCTGAR QVITLLNELK RRGKRAYGVV SMCIGTGMGA AAVFEYPGN
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name ACAA1
Conjugate Unconjugated
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.