Learn More
Novus Biologicals™ APO-BRDU-IHC (TUNEL) Apoptosis Kit
SDP
Catalog No. NBP231164 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
NBP231164 1 Kit
1 options
Catalog No. NBP231164 Supplier Novus Biologicals™ Supplier No. NBP231164
Add to Cart
Add to Cart
Applications: Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Specifications
For Use With (Application) Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin
Product Type APO-BRDU-IHC (TUNEL) Apoptosis Kit
Content And Storage Positive and Negative Control Slides, Blocking Buffer, H202/Urea Tablets, Proteinase K, DAB Tablets, TdT Enzyme, Br-dUTP, 200X Conjugate, 5X-Reaction Buffer, Biotin-anti BrdU antibody, Methyl Green. Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 1 Kit