Learn More
Description
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 0.1M NaCl, 1mM DTT |
| Quantity | 250 μg |
| Source | Human |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | ARL4 |
| Conjugate | Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
