Learn More
Description
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | Dot Blot, SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 20% glycerol, 0.2mM PMSF |
| Molecular Weight (g/mol) | 23.3kDa |
| Purification Method | SDS-PAGE |
| Quantity | 0.05 mg |
| Source | Human |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | ARL5A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
