Learn More
Novus Biologicals™ Recombinant E. coli Disulfide oxidoreductase Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris buffer (pH 7.5), 2mM EDTA |
| Gene ID (Entrez) | 948353 |
| Molecular Weight (g/mol) | 21.1kDa |
| Name | Disulfide oxidoreductase Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Immunogen | AQYEDGKQYTTLEKPVAGAPQVLEFFSFFCPHCYQFEEVLHISDNVKKKLPEGVKMTKYHVNFMGGDLGKDLTQAWAVAMALGVEDKVTVPLFEGVQKTQTIRSASDIRDVFINAGIKGEEYDAAWNSFVVKSLVAQQEKAAADVQLRGVPAMFVNGKYQLNPQGMDTSNMDVFVQQYADTVKYLSEKK |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Bacteria |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.