Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant E. coli Ferric uptake regulator Protein
SDP

Catalog No. NBC118429 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118429 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118429 Supplier Novus Biologicals™ Supplier No. NBC118429
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-148 of E.coli Ferric uptake regulator The Recombinant E. coli Ferric uptake regulator Protein is derived from E. coli. The Recombinant E. coli Ferric uptake regulator Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 2 mM CaCl2, 100 mM NaCl
Molecular Weight (g/mol) M.W. (theoretical): 16.7 kDa
Name Ferric uptake regulator Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MTDNNTALKKAGLKVTLPRLKILEVLQEPDNHHVSAEDLYKRLIDMGEEIGLATVYRVLNQFDDAGIVTRHNFEGGKSVFELTQQHHHDHLICLDCGKVIEFSDDSIEARQREIAAKHGIRLTNHSLYLYGHCAEGDCREDEHAHEGK
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Symbol fur
Cross Reactivity Bacteria
Research Category Cell Cycle and Replication
Purity or Quality Grade >95%, by SDS-PAGE
Protein Ferric uptake regulator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.