Learn More
Novus Biologicals™ beta Galactosidase IHC Kit
SDP
Catalog No. nbp229546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
NBP229546 1 Kit
1 options
Catalog No. NBP229546 Supplier Novus Biologicals™ Supplier No. NBP229546
beta Galactosidase IHC Kit
Specifications
Content And Storage Storage is content dependent.
Quantity 1 Kit
Kit Contents Fixative Solution (25% Glutaraldehyde) 1mL, X-Gal (40 mg/ml in DMSO) 600μL, Staining Solution A (0.1M potassium ferricyanide) 10mL, Staining Solution B (0.1M potassium ferrocyanide) 10mL, Staining Solution C (1M MgCl2) 1mL
Product Type beta Galactosidase IHC Kit