Learn More
Description
Specifications
Specifications
| Species | E. coli |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-76322. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | EC 3.2.1.23; b-gal |
| Product Type | beta Galactosidase |
| Quantity | 0.05 mg |
| Regulatory Status | RUO |
| Source | Synthetic |
| Termination | 17 amino acids near the amino terminus of E coli beta-galactosidase |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
