Learn More
Novus Biologicals™ Blood Leukocyte Whole Cell Lysate

Description
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Blood Lymph |
| Quantity | 1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Total Protein - Human Adult Normal Tissue: Peripheral Blood Leukocyte |
| Cellular Fraction | Whole |
| Host Species | Human |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.