Learn More
Description
Specifications
Specifications
| Antigen | IgG, Fc Protein |
| Applications | Western Blot, ELISA, Dot Blot |
| Dilution | Western Blot, ELISA, Dot Blot, SDS-Page |
| Formulation | Potassium Phosphate, 0.15M Sodium Chloride pH 7.2. |
| Quantity | 1 mg |
| Content And Storage | Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
