Learn More
R&D Systems™ Carbonic Anhydrase IX/CA9 Antibody Blocking Peptide
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Gene ID (Entrez) | 768 |
| Species | Human |
| Product Line | Novus |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB 100-417. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58 N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 |
| Gene Symbol | CA9 |
| Product Type | Carbonic Anhydrase IX/CA9 |
| Quantity | 0.05 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.