Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Casein Kinase 2 alpha His Protein
SDP

Catalog No. NBC118386 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118386 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118386 Supplier Novus Biologicals™ Supplier No. NBC118386
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-391 of Human Casein Kinase 2 alpha The Recombinant Human Casein Kinase 2 alpha Protein is derived from E. coli. The Recombinant Human Casein Kinase 2 alpha Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001886
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 500 mM NaCl, 1 mM DTT, 50% glycerol
Gene ID (Entrez) 1457
Molecular Weight (g/mol) M.W. (theoretical): 47.3 kDa
Name Casein Kinase 2 alpha protein
Purification Method Protein
Quantity 0.05 mg
Source E. Coli
Immunogen 1-391aa Human, Recombinant, His-tagged, E.coli
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol CSNK2A1
Conjugate Unconjugated
Recombinant Recombinant
Research Category DNA Repair, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Purity or Quality Grade >90%, by SDS-PAGE
Protein Casein Kinase 2 alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.