Learn More
Specifications
Specifications
| For Use With (Application) | Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin |
| Product Type | Endoplasmic Reticulum Enrichment Extraction Kit |
| Content And Storage | 5X Isosmotic Homogenization Buffer (HEPES Sucrose KCL), 1X Suspension Buffer (HEPES Sucrose KCL EGTA), 10X PBS, 5X Calcium Chloride Solution (CaCl2), 00X Protease Inhibitor Cocktail (PIC). Storage is content dependent. |
| Product Line | Novus |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
