Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ETS1 associated protein II Antibody Blocking Peptide
SDP

Catalog No. NBP211491E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211491E 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211491E Supplier Novus Biologicals™ Supplier No. NBP211491PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Synthetic peptide corresponding to amino acids 330-360 of human Tyrosyl-DNA phosphodiesterase protein. The ETS1 associated protein II Blocking Peptide is derived from Synthetic. The ETS1 associated protein II Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 51567
Species Human
Purity >85%, by HPLC
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation Sodium Bicarbonate buffer, pH 8.00 containing up to 10% DMSO
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP2-11491. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5 '-Tyr-DNA phosphodiesterase, MGC9099,5 '-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2
Gene Symbol TDP2
Product Type ETS1 associated protein II
Quantity 0.25 mg
Regulatory Status RUO
Termination Blocking Peptide for NBP2-11491
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.