Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FABP2/I-FABP Antibody Blocking Peptide
SDP

Catalog No. NB10059746E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10059746E 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10059746E Supplier Novus Biologicals™ Supplier No. NB10059746PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide from bovine FABP2. Source: Synthetic Amino Acid Sequence: C-EGVEAKRIFKKD The FABP2/I-FABP Blocking Peptide is derived from Synthetic. The FABP2/I-FABP Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 2169
Species Bovine
Purity >50%, by HPLC
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-59746 PEP. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein
Gene Symbol FABP2
Product Type FABP2/I-FABP
Quantity 0.1 mg
Regulatory Status RUO - research use only
Termination C-EGVEAKRIFKKD
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.