Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FBXW11 Antibody Blocking Peptide
SDP

Catalog No. NBP196762 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196762 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196762 Supplier Novus Biologicals™ Supplier No. NBP196762
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide from FBXW11. Source: Synthetic The FBXW11 Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 23291
Species Human
Purity >95%
Accession Number Q9UKB1
Content And Storage Store at -20 C short term. Aliquot and store (at -80 C long term. Avoid freeze-thaw cycles.)
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
For Use With (Application) This product should be used at 1.0μg per 1.0μL of antiserum in per assay.
Gene Alias beta-transducin repeat-containing protein 2, BTRC2F-box/WD repeat-containing protein 1 B, BTRCP2FBW1B, F-box and WD repeat domain containing 11, F-box and WD-40 domain protein 11, F-box and WD-40 domain protein 1 B, F-box protein Fbw1b, F-box/WD repeat-containing protein 11, Fbw11, FBXW1BFbw1b, Homologous to Slimb protein, Hos, KIAA0696F-box and WD repeats protein beta-TrCP2
Gene Symbol FBXW11
Product Type FBXW11 Peptide
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Termination Sequence: Leu - Val - Phe - Phe - Ala - Glu - Asp - Val - Gly - Ser - Asn - Lys - OH (aa 17-28)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.