Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ GALK1 Protein
SDP

Catalog No. NBP1452945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1452945 0.5 mg
NBP14529401 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1452945 Supplier Novus Biologicals™ Supplier No. NBP1452940.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-392 of Human GALK1 The Recombinant Human GALK1 Protein is derived from E. coli. The Recombinant Human GALK1 Protein has been validated for the following applications: Western Blot, SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 0.1M NaCl, 1mM DTT
Gene ID (Entrez) 2584
Molecular Weight (g/mol) 44.4kDa
Purification Method Protein
Quantity 0.5 mg
Source Human
Immunogen MGSSHHHHHH SSGLVPRGSH MAALRQPQVA ELLAEARRAF REEFGAEPEL AVSAPGRVNL IGEHTDYNQG LVLPMALELM TVLVGSPRKD GLVSLLTTSE GADEPQRLQF PLPTAQRSLE PGTPRWANYV KGVIQYYPAA PLPGFSAVVV SSVPLGGGLS SSASLEVATY TFLQQLCPDS GTIAARAQVC QQAEHSFAGM PCGIMDQFIS LMGQKGHALL IDCRSLETSL VPLSDPKLAV LITNSNVRHS LASSEYPVRR RQCEEVARAL GKESLREVQL EELEAARDLV SKEGFRRARH VVGEIRRTAQ AAAALRRGDY RAFGRLMVES HRSLRDDYEV SCPELDQLVE AALAVPGVYG SRMTGGGFGG CTVTLLEASA APHAMRHIQE HYGGTATFYL SQAADGAKVL CL
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name GALK1
Conjugate Unconjugated
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.