Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ gp96/HSP90B1/GRP94 Protein
SDP

Catalog No. NBC12105810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC12105810 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC12105810 Supplier Novus Biologicals™ Supplier No. NBC121058100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 22-803 of Human gp96/HSP90B1/GRP94 The Recombinant Human gp96/HSP90B1/GRP94 Protein is derived from E. coli. The Recombinant Human gp96/HSP90B1/GRP94 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003290
Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 1mM EDTA, 0.1M NaCl, 10% glycerol
Gene ID (Entrez) 7184
Molecular Weight (g/mol) 94.4kDa
Purification Method Affinity purified
Quantity 100 μg
Source Recombinant human GRP94 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen Recombinant human GRP94 (HSP90B1), 22-803 aa (NP_003290), fused to His-tag at N-terminus, was expressed in E. coli and purified by using conventional chromatography
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name gp96/HSP90B1/GRP94
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.