Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ GSTT1 Protein
SDP

Catalog No. NBC12878201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC12878201 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC12878201 Supplier Novus Biologicals™ Supplier No. NBC1287820.1MG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-240 of Human GSTT1 The Recombinant Human GSTT1 Protein is derived from E. coli. The Recombinant Human GSTT1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number AAH07065
Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol
Gene ID (Entrez) 2952
Molecular Weight (g/mol) 31.5kDa
Purification Method Protein
Quantity 0.1 mg
Source Recombinant human GSTT1, fused to His-tag at N-terminus, expressed in E. coli
Immunogen GSTT1, 1-240aa, Human, His-tagged, Recombinant, E. coli (AAH07065).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name GSTT1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.