Learn More
Novus Biologicals™ HL60 Cell Lysate
SDP
Catalog No. NBP225052
Change view
Click to view available options
Quantity:
0.1 mg
Catalog No. Quantity
NBP225052 0.1 mg
1 options
Catalog No. NBP225052 Supplier Novus Biologicals™ Supplier No. NBP225052
Customers require a valid business account associated to their profile to access pricing and ordering options.
The HL60 Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.
Specifications
Cell Line HL6 cell line lysate (promyelocytic cell line)
Quantity 0.1 mg
Storage Buffer 1X SDS sample buffer: 60mM Tris-HCl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol
Content And Storage -80°C in the dark. Avoid freeze-thaw cycles.